Fmoc-trans-4-fluoro-Pro-Oh

Supplier: BACHEM AMERICAS INC

Synonyms: (2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-fluoropyrrolidine-2-carboxylic acid, (2S,4R)-4-Fluoropyrrolidine-2-carboxylic acid, N-FMOC protected, Fmoc-trans-4-F-Pro-OH

4034299.0250 4034299.1000 4034299.5000
F-4035.0250BAEA 478.3 CAD
F-4035.0250BA F-4035.1000BA F-4035.5000BA
Fmoc-trans-4-fluoro-Pro-Oh
Fmoc-trans-4-fluoro-Pro-Oh

Sequence: Fmoc-trans-4-fluoro-Pro-OH


Flp derivatives.


Synonym(s): (2S,4R)-4-Fluoro-1-Fmoc-pyrrolidine-2-carboxylic acid

Formula: C₂₀H₁₈FNO₄
Storage Temperature: Freezer
MDL Number: MFCD03424224
CAS Number: 203866-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR