trans-4-Fluoro-L-proline

Supplier: BACHEM AMERICAS INC

4033275.0250 4033275.1000 4033275.5000
F-3975.0250BAEA 464.74 CAD
F-3975.0250BA F-3975.1000BA F-3975.5000BA
trans-4-Fluoro-L-proline
trans-4-Fluoro-L-proline

Sequence: H-trans-4-Fluoro-Pro-OH


Flp derivatives.


Synonym(s): (2S,4R)-4-Fluoro-pyrrolidine-2-carboxylic acid

Formula: C₅H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD02683982
CAS Number: 2507-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR