(2S,4S)-4-Fluoropyrrolidine-2-carboxylic acid

Supplier: BACHEM AMERICAS INC

Synonyms: H-cis-4-Fluoro-Pro-OH

4033274.0250 4033274.1000 4033274.5000
F-3970.0250BAEA 309.81 CAD
F-3970.0250BA F-3970.1000BA F-3970.5000BA
(2S,4S)-4-Fluoropyrrolidine-2-carboxylic acid
(2S,4S)-4-Fluoropyrrolidine-2-carboxylic acid

Sequence: H-cis-4-Fluoro-Pro-OH


Flp derivatives.


Synonym(s): (2S,4S)-4-Fluoro-pyrrolidine-2-carboxylic acid

Formula: C₅H₈FNO₂
MW: 133.122 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02683983
CAS Number: 2438-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR