Phenylacetylglutamine

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-5-Amino-5-oxo-2-(2-phenylacetamido)pentanoic acid, Phenylac-Gln-OH

4029485.0001 4029485.0005
F-3955.0001BAEA 185.3 CAD
F-3955.0001BA F-3955.0005BA
Phenylacetylglutamine
Phenylacetylglutamine

Sequence: Phenylac-Gln-OH


For isoglutamine derivatives please see the family "Glutamic Acid".


Phenylacetyl-L-glutamine is formed in the liver of primates and humans by condensation of glutamine with phenylacetyl-CoA and can be detected by HPLC or GC-MS in the urine. Furthermore, it is a component of 3-phenylacylamino-2,6-piperidinone (PAP), a naturally occurring peptide derivative used for inhibiting growth of cancer tissues, and can be used as reference compound in the enantioseparation of PAP.

Formula: C₁₃H₁₆N₂O₄
MW: 264.281 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01725188
CAS Number: 28047-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR