2-Aminobutanamide

Supplier: BACHEM AMERICAS INC

Synonyms: H-Abu-NH₂, (S)-2-Aminobutanamide

4029269.0001 4029269.0005
F-3925.0001BAEA 309.81 CAD
F-3925.0001BA F-3925.0005BA
2-Aminobutanamide
2-Aminobutanamide

Sequence: H-Abu-NH₂

Formula: C₄H₁₀N₂O
MW: 102.137 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00136552
CAS Number: 7324-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR