H-D-Asp-OtBu

Supplier: BACHEM AMERICAS INC

4028966.0001 4028966.0005
F-3855.0001BAEA 516.34 CAD
F-3855.0001BA F-3855.0005BA
H-D-Asp-OtBu
H-D-Asp-OtBu

Sequence: H-D-Asp-OtBu


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₈H₁₅NO₄
Storage Temperature: Freezer
MDL Number: MFCD00270618
CAS Number: 148823-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR