γ-Aminobutyric acid-t-butyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

4028489.0001 4028489.0005
F-3755.0001BAEA 185.46 CAD
F-3755.0001BA F-3755.0005BA
γ-Aminobutyric acid-t-butyl ester hydrochloride
gamma-Aminobutyric acid-t-butyl ester hydrochloride

Sequence: H-γ-Abu-OtBu · HCl


Flexible spacer molecules of various length.


Synonym(s): H-GABA-OtBu · HCl

Formula: C₈H₁₈ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00270592
CAS Number: 58640-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR