N-Me-D-Tyr-OH
Supplier: BACHEM AMERICAS INC
4028208.0250
4028208.1000
F-3715.0250BAEA
439.32
CAD
F-3715.0250BA
F-3715.1000BA
N-Me-D-Tyr-OH
N-Me-D-Tyr-OH
Sequence: N-Me-D-Tyr-OH
CAS-No.: 178357-84-1
Mol.weight: 195.22
SumFormula: C₁₀H₁₃NO₃
Bulk item no.: F-3715
Synonym(s):
EINECS no.: -
MDL no.: MFCD02094309
Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
Formula:
C₁₀H₁₃NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD02094309 CAS Number: 178357-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...