H-β,β-Dicyclohexyl-DL-Ala-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
H-β,β-Dicyclohexyl-DL-Ala-OH
4028091.0250
4028091.1000
F-3695.0250BAEA
877.75
CAD
F-3695.0250BA
F-3695.1000BA
H-β,β-Dicyclohexyl-DL-Ala-OH
H-beta,beta-Dicyclohexyl-DL-Ala-OH
Sequence: H-β,β-Dicyclohexyl-DL-Ala-OH
Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines
Dip derivatives.
Formula:
C₁₅H₂₇NO₂ MW: 253.386 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD02259524 CAS Number: 274262-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...