N-ω-Hydroxy-L-norarginine acetate salt

Supplier: BACHEM AMERICAS INC

4027934.0050 4027934.0250 4027934.1000
F-3685.0050BAEA 264.68 CAD
F-3685.0050BA F-3685.0250BA F-3685.1000BA
N-ω-Hydroxy-L-norarginine acetate salt
N-ω-Hydroxy-L-norarginine acetate salt

Sequence: N-ω-Hydroxy-L-norarginine


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).


Synonym(s): L-2-Amino-4-(2'-hydroxyguanidino)-butyric acid#nor-NOHA#H-Nar(OH)-OH

Formula: C₅H₁₂N₄O₃.CH₃COOH
Storage Temperature: Freezer
MDL Number: MFCD01318777
CAS Number: 189302-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR