3-Amino-L-tyrosine dihydrochloride
Supplier: BACHEM AMERICAS INC
Synonyms:
3-NH2-Tyr-OH·2HCl
, H-Tyr(3-NH2)-OH·2HCl
, H-3-Amino-Tyr-OH · 2 HCl
4027898.0005
4027898.0001
F-3675.0005BAEA
397.32
CAD
F-3675.0005BA
F-3675.0001BA
3-Amino-L-tyrosine dihydrochloride
3-Amino-L-tyrosine dihydrochloride
Sequence: H-3-Amino-Tyr-OH · 2 HCl
Bachem offers derivatives of (di)halogenated tyrosines, (di)nitrotyrosine and other 3-substituted tyrosines.
See also 3-Methoxytyramine · HCl (Q-1330).
Formula:
C₉H₁₂N₂O₃·2HCl MW: 269.13 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00012964 CAS Number: 23279-22-3 EINECS: 245-552-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...