Albizziin

Supplier: BACHEM AMERICAS INC

Synonyms: L-Albizziine

4027854.0250 4027854.1000
F-3670.0250BAEA 1918.25 CAD
F-3670.0250BA F-3670.1000BA
Albizziin
Albizziin

Sequence: H-β-(Ureido)-Ala-OH


Abbreviated Dap, the abbreviation Dpr can be found in the literature as well. See also Albiziine, Nβ-carbamoyl-Dab, F-3670.


Synonym(s): H-β-((Aminocarbonyl)amino)-Ala-OH#H-Dap(carbamoyl)-OH

Formula: C₄H₉N₃O₃
MW: 147.134 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00007952
CAS Number: 1483-07-4
EINECS: 216-046-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR