Spinacine

Supplier: BACHEM AMERICAS INC

Synonyms: L-4,5,6,7-Tetrahydro-1H-imidazo[4,5-c]pyridine-6-carboxylic acid

4027853.0001 4027853.0005
F-3665.0001BAEA 291.24 CAD
F-3665.0001BA F-3665.0005BA
Spinacine
Spinacine

Sequence: L-4,5,6,7-Tetrahydro-1H-imidazo[4,5-c]pyridine-6-carboxylic acid


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): Spinacine

Formula: C₇H₉N₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD01318776
CAS Number: 59981-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR