L-4-Cyanophenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: 4-Cyano-L-phenylalanine, H-Phe(4-CN)-OH, (2S)-2-Amino-3-(4-cyanophenyl)propanoic acid

4028063.0001 4028063.0005
F-3610.0001BAEA 903.62 CAD
F-3610.0001BA F-3610.0005BA
L-4-Cyanophenylalanine
L-4-Cyanophenylalanine

Sequence: H-4-Cyano-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.


Key intermediate for the synthesis of amidines, which can be used as arginine mimetics. McMurray obtained p-Tetrazolylphenylalanine by 1,3-dipolar cycloaddition with trimethyltin azide. Additionally, p-Cyanophenylalanine may serve as well as a IR- or Raman-probe in peptides and recombinant proteins. As a fluorophore, it has been used in combination with Trp in FRET substrates.

Formula: C₁₀H₁₀N₂O₂
MW: 190.202 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00270363
CAS Number: 167479-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR