H-P-Carboxy-PHE(OtBu)-OH

Supplier: BACHEM AMERICAS INC

4027709.0001 4027709.0005
F-3585.0001BAEA 1135.93 CAD
F-3585.0001BA F-3585.0005BA
H-P-Carboxy-PHE(OtBu)-OH
H-P-Carboxy-PHE(OtBu)-OH

Sequence: H-p-Carboxy-Phe(OtBu)-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.

Formula: C₁₄H₁₉NO₄
Storage Temperature: Freezer
MDL Number: MFCD01318767
CAS Number: 218962-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR