N-Me-DL-Phe-OMe·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: N-Me-DL-Phe-OMe·HCl

4027307.0001 4027307.0005
F-3545.0001BAEA 516.15 CAD
F-3545.0001BA F-3545.0005BA
N-Me-DL-Phe-OMe·HCl
N-Me-DL-Phe-OMe·HCl

Sequence: N-Me-DL-Phe-OMe · HCl


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
Short N-methyl-phenylalanine-rich peptides were shown to function as versatile blood-brain barrier shuttles.


This building block has been used in the synthesis of 1,4-benzodiazepine-2,5-diones.

Formula: C₁₁H₁₆ClNO₂
MW: 229.704 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08458660
CAS Number: 16975-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR