(S)-2-amino-2,3-dimethylbutanoic acid
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-2-Amino-2,3-dimethylbutanoic acid, H-α-Me-Val-OH
4026440.0001
4026440.0005
F-3535.0001BAEA
227.28
CAD
F-3535.0001BA
F-3535.0005BA
(S)-2-amino-2,3-dimethylbutanoic acid
(S)-2-amino-2,3-dimethylbutanoic acid
Sequence: H-α-Me-Val-OH
Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.
MeVal, αMeVal.
Formula:
C₆H₁₃NO₂ MW: 131.175 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01318764 CAS Number: 53940-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...