L-Cyclopropylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-Amino-3-cyclopropylpropanoic acid, H-Cyclopropyl-Ala-OH, H-β-Cyclopropyl-Ala-OH

4026826.0001 4026826.0005
F-3470.0001BAEA 653.27 CAD
F-3470.0001BA F-3470.0005BA
L-Cyclopropylalanine
L-Cyclopropylalanine

Sequence: H-β-Cyclopropyl-Ala-OH


Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines


β-Cyclopropylalanine has been isolated in 1986 from Amanita virgineoides Bas., and was shown to inhibit spore germination of Pyricularia oryzae Cav., the causative agent of rice blast disease. This unusual amino acid was found to be a potent antagonist to E.coli ATCC 9723.

Formula: C₆H₁₁NO₂
Storage Temperature: Freezer
MDL Number: MFCD00798687
CAS Number: 102735-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR