(R)-Methyl-2-amino-3-chloropropanoate hydrochloride
Supplier: BACHEM AMERICAS INC
Synonyms:
(R)-Methyl 2-amino-3-chloropropanoate hydrochloride, H-β-Chloro-Ala-OMe·HCl
4002402.0001
4002402.0005
F-3465.0001BAEA
283.94
CAD
F-3465.0001BA
F-3465.0005BA
(R)-Methyl-2-amino-3-chloropropanoate hydrochloride
(R)-Methyl-2-amino-3-chloropropanoate hydrochloride
Sequence: H-β-Chloro-Ala-OMe · HCl
Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines
CAS Number (net): 740771-74-8 .
Formula:
C₄H₉Cl₂NO₂ MW: 174.021 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00270526 CAS Number: 17136-54-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...