H-L-Dap(boc)-OMe·HCl

Supplier: BACHEM AMERICAS INC

4026562.0001 4026562.0005 4026562.0025
F-3420.0001BAEA 335.58 CAD
F-3420.0001BA F-3420.0005BA F-3420.0025BA
H-L-Dap(boc)-OMe·HCl
H-L-Dap(boc)-OMe·HCl

Sequence: H-Dap(Boc)-OMe · HCl


Abbreviated Dap, the abbreviation Dpr can be found in the literature as well. See also Albiziine, Nβ-carbamoyl-Dab, F-3670.

Formula: C₉H₁₉ClN₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00798684
CAS Number: 114559-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR