Serine O-sulfate

Supplier: BACHEM AMERICAS INC

4026389.0001 4026389.0005
F-3365.0001BAEA 309.66 CAD
F-3365.0001BA F-3365.0005BA
Serine O-sulfate
Serine O-sulfate

Sequence: H-Ser(SO₃H)-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.
N-Terminal phosphoserine residues react with thioesters yielding highly reactive acylphosphates which spontaneously undergo an intramolecular O-N-shift allowing chemical ligation at such positions.

Formula: C₃H₇NO₆S
Storage Temperature: Freezer
MDL Number: MFCD00672378
CAS Number: 626-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR