N-(5-Amino-5-carboxypentyl)acetamidine

Supplier: BACHEM AMERICAS INC

Synonyms: H-Lys(acetimidoyl)-OH

4026182.0001 4026182.0005
F-3325.0001BAEA 993.74 CAD
F-3325.0001BA F-3325.0005BA
N-(5-Amino-5-carboxypentyl)acetamidine
N-(5-Amino-5-carboxypentyl)acetamidine

Sequence: H-Lys(acetimidoyl)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
See also Z-Lys-ONp · HCl, M-1295.


Synonym(s): L-NIL#H-Lys(1-iminoethyl)-OH#N-(5-Amino-5-carboxypentyl)-acetamidine

Formula: C₈H₁₇N₃O₂
MW: 187.243 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00792818
CAS Number: 53774-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR