H-β-(1,2,4-Triazol-1-yl)-DL-Ala-OH

Supplier: BACHEM AMERICAS INC

4025514.0001 4025514.0005
F-3225.0001BAEA 1616.07 CAD
F-3225.0001BA F-3225.0005BA
H-β-(1,2,4-Triazol-1-yl)-DL-Ala-OH
H-β-(1,2,4-Triazol-1-yl)-DL-Ala-OH

Sequence: H-β-(1,2,4-Triazol-1-yl)-DL-Ala-OH


Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines


Triazolylalanine is a common metabolite of triazole fungicides and pesticides in plants.

Formula: C₅H₈N₄O₂
Storage Temperature: Freezer
MDL Number: MFCD00077168
CAS Number: 86362-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR