L-Threoninol

Supplier: BACHEM AMERICAS INC

4025486.0001 4025486.0005
F-3200.0001BAEA 387.21 CAD
F-3200.0001BA F-3200.0005BA
L-Threoninol
L-Threoninol

Sequence: L-Threoninol

Formula: C₄H₁₁NO₂
Storage Temperature: Freezer
MDL Number: MFCD00191173
CAS Number: 3228-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR