3-(1-tritylimidazol-4-yl)propanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: N-1-Trityl-deamino-histidine

4025149.0001 4025149.0005
F-3190.0001BAEA 645.43 CAD
F-3190.0001BA F-3190.0005BA
3-(1-tritylimidazol-4-yl)propanoic acid
3-(1-tritylimidazol-4-yl)propanoic acid

Sequence: N-1-Trityl-deamino-histidine


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): N-1-Trityl-dihydrourocanic acid#3-(N-1-Trityl-imidazol-4-yl)-propionic acid

Formula: C₂₅H₂₂N₂O₂
MW: 382.463 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237290
CAS Number: 160446-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR