3-(1H-Imidazol-4-yl)propanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: Deamino-histidine, Imidazole-5-propionic acid, Imidazolepropionic acid

4025148.0001 4025148.0005
F-3185.0001BAEA 516.34 CAD
F-3185.0001BA F-3185.0005BA
3-(1H-Imidazol-4-yl)propanoic acid
3-(1H-Imidazol-4-yl)propanoic acid

Sequence: Deamino-histidine


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): Dihydrourocanic acid#3-(Imidazol-4-yl)-propionic acid

Formula: C₆H₈N₂O₂
MW: 140.142 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237415
CAS Number: 1074-59-5
EINECS: 214-045-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR