H-D-Glu(OtBu)-OtBu·HCl
Supplier: BACHEM AMERICAS INC
Synonyms:
H-D-Glu(OtBu)-OtBu·HCl, (R)-Di-tert-butyl 2-aminopentanedioate hydrochloride
4025307.0001
4025307.0005
F-3165.0001BAEA
232.35
CAD
F-3165.0001BA
F-3165.0005BA
H-D-Glu(OtBu)-OtBu·HCl
H-D-Glu(OtBu)-OtBu·HCl
Sequence: H-D-Glu(OtBu)-OtBu · HCl
The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
Formula:
C₁₃H₂₆ClNO₄ MW: 295.804 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00237068 CAS Number: 172793-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...