H-AAD(OtBu)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-Amino-6-(tert-butoxy)-6-oxohexanoic acid

4025236.0001 4025236.0005
F-3150.0001BAEA 903.62 CAD
F-3150.0001BA F-3150.0005BA
H-AAD(OtBu)-OH
H-AAD(OtBu)-OH

Sequence: H-Aad(OtBu)-OH
Synonym(s): L-α-Aminoadipic acid-δ-t-butyl ester#(S)-2-Amino-hexandioic acid-6-t-butyl ester#H-Homoglu(OtBu)-OH

Formula: C₁₀H₁₉NO₄
MW: 217.265 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08272285
CAS Number: 201354-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR