H-D-Asp(OMe)-OH

Supplier: BACHEM AMERICAS INC

4025294.0001 4025294.0005
F-3135.0001BAEA 309.81 CAD
F-3135.0001BA F-3135.0005BA
H-D-Asp(OMe)-OH
H-D-Asp(OMe)-OH

Sequence: H-D-Asp(OMe)-OH · HCl


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₅H₉NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD06797583
CAS Number: 21394-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR