(R)-2,3-Diaminopropanoic acid hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: 3-Amino-D-alanine Hydrochloride, H-D-Dap-OH.HCl, H-D-Dap-OH·HCl

4025043.0001 4025043.0005
F-3045.0001BAEA 380.91 CAD
F-3045.0001BA F-3045.0005BA
(R)-2,3-Diaminopropanoic acid hydrochloride
(R)-2,3-Diaminopropanoic acid hydrochloride

Sequence: H-D-Dap-OH · HCl


Abbreviated Dap, the abbreviation Dpr can be found in the literature as well. See also Albiziine, Nβ-carbamoyl-Dab, F-3670.


Growth inhibitor for microorganisms.

Formula: C₃H₉ClN₂O₂
MW: 140.5688 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00070606
CAS Number: 6018-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR