trans-4-Hydroxy-D-proline

Supplier: BACHEM AMERICAS INC

4025355.0001 4025355.0005
F-2980.0001BAEA 1041.57 CAD
F-2980.0001BA F-2980.0005BA
trans-4-Hydroxy-D-proline
trans-4-Hydroxy-D-proline

Sequence: H-D-Hyp-OH


Hyp denotes L-trans-hydroxyproline.


Synonym(s): (2R,4S)-4-Hydroxypyrrolidine-2-carboxylic acid

Formula: C₅H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00065608
CAS Number: 3398-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR