2,6-Diaminohex-4-enoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: H-trans-4,5-Dehydro-DL-Lys-OH·2 HCl

4017146.0250 4017146.1000
F-2970.0250BAEA 593.81 CAD
F-2970.0250BA F-2970.1000BA
2,6-Diaminohex-4-enoic acid
2,6-Diaminohex-4-enoic acid

Sequence: H-trans-4,5-Dehydro-DL-Lys-OH · 2 HCl


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


Synonym(s): DL-trans-2,6-Diamino-4-hexenoic acid · 2 HCl

Formula: C₆H₁₂N₂O₂
MW: 144.174 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08271758
CAS Number: 39871-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR