L-Methionine sulfoxide

Supplier: BACHEM AMERICAS INC

4014135.0005 4014135.0025
F-2945.0005BAEA 180.91 CAD
F-2945.0005BA F-2945.0025BA
L-Methionine sulfoxide
L-Methionine sulphoxide

Sequence: H-Met(O)-OH


Oxidation of Met to the sulfoxide Met(O) can be easily reverted, whereas sulfone (Met(O₂)) formation is practically irreversible.


(Diastereomeric mixture)

Formula: C₅H₁₁NO₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00063093
CAS Number: 3226-65-1
EINECS: 221-758-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR