(R)-2-Amino-3-(4-benzoylphenyl)propanoic acid

Supplier: BACHEM AMERICAS INC

4017834.0001 4017834.0005
F-2810.0001BAEA 439.41 CAD
F-2810.0001BA F-2810.0005BA
(R)-2-Amino-3-(4-benzoylphenyl)propanoic acid
(R)-2-Amino-3-(4-benzoylphenyl)propanoic acid

Sequence: H-p-Bz-D-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.
4-Bpa is incorporated into peptides as a photolabel or for photocrosslinking.


Synonym(s): H-D-Bpa-OH

Formula: C₁₆H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00238101
CAS Number: 201466-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR