L-4-Benzoylphenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-Amino-3-(4-benzoylphenyl)propanoic acid, H-Bpa-OH, H-p-Bz-Phe-OH, H-Phe(4-Bz)-OH

4017646.0001 4017646.0005
F-2800.0001BAEA 462.56 CAD
F-2800.0001BA F-2800.0005BA
L-4-Benzoylphenylalanine
L-4-Benzoylphenylalanine

Sequence: H-p-Bz-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.
4-Bpa is incorporated into peptides as a photolabel or for photocrosslinking.


Synonym(s): H-Bpa-OH

Formula: C₁₆H₁₅NO₃
MW: 269.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00238102
CAS Number: 104504-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR