L-Homoarginine

Supplier: BACHEM AMERICAS INC

4016423.0005 4016423.0025
F-2780.0005BAEA 317.74 CAD
F-2780.0005BA F-2780.0025BA
L-Homoarginine
L-Homoarginine

Sequence: H-Homoarg-OH


Har derivatives.


Synonym(s): L-α-Amino-ε-guanidinohexanoic acid

Formula: C₇H₁₆N₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00237108
CAS Number: 156-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR