(S)-2-Aminooctanedioic acid

Supplier: BACHEM AMERICAS INC

4017100.0001 4017100.0005
F-2740.0001BAEA 1187.51 CAD
F-2740.0001BA F-2740.0005BA
(S)-2-Aminooctanedioic acid
(S)-2-Aminooctanedioic acid

Sequence: H-Asu-OH
Synonym(s): (S)-2-Aminooctanedioic acid#L-α-Aminosuberic acid

Formula: C₈H₁₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00037224
CAS Number: 4254-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR