H-Lys(Me)₃-OH chloride

Supplier: BACHEM AMERICAS INC

4016650.0050 4016650.0250
F-2665.0050BAEA 232.35 CAD
F-2665.0050BA F-2665.0250BA
H-Lys(Me)₃-OH chloride
H-Lys(Me)₃-OH chloride

Sequence: H-Lys(Me)₃-OH chloride


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₉H₂₁ClN₂O₂
Storage Temperature: Freezer
MDL Number: MFCD02259519
CAS Number: 55528-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR