Pam3Cys-OH

Supplier: BACHEM AMERICAS INC

4015000.0250 4015000.1000
F-2630.0250BAEA 1384.92 CAD
F-2630.0250BA F-2630.1000BA
Pam3Cys-OH
Pam3Cys-OH

Sequence: Palmitoyl-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-OH
Synonym(s): Pam₃Cys-OH


The lipophilic cysteine derivative Pam₃Cys resembles the N-terminus of the lipoprotein from E.coli outer membranes. Pam₃Cys has been successfully used in the synthesis of immunogenic conjugates like peptide mitogens or vaccines. For coupling protocols see the papers of Zeng et al.

Formula: C₅₄H₁₀₃NO₇S
Storage Temperature: Freezer
MDL Number: MFCD00153468
CAS Number: 87420-41-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR