Myristoyl-Gly-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
2-Tetradecanamidoacetic acid
4015914.0050
4015914.0250
F-2550.0050BAEA
309.81
CAD
F-2550.0050BA
F-2550.0250BA
Myristoyl-Gly-OH
Myristoyl-Gly-OH
Sequence: Myristoyl-Gly-OH
Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.
Formula:
C₁₆H₃₁NO₃ MW: 285.428 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00155627 CAS Number: 14246-55-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...