(2S,3S)-Methyl-2-amino-3-hydroxybutanoate hydrochloride

Supplier: BACHEM AMERICAS INC

4016116.0001 4016116.0005
F-2545.0001BAEA 413.58 CAD
F-2545.0001BA F-2545.0005BA
(2S,3S)-Methyl-2-amino-3-hydroxybutanoate hydrochloride
(2S,3S)-Methyl-2-amino-3-hydroxybutanoate hydrochloride

Sequence: H-allo-Thr-OMe · HCl


Bachem also offers phosphothreonine derivatives.
aThr derivatives.

Formula: C₅H₁₂ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00237766
CAS Number: 79617-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR