Bz-Cit-OMe
Supplier: BACHEM AMERICAS INC
Synonyms:
Benzoyl-L-citrulline methyl ester, Bz-Orn(carbamoyl)-OMe
4014973.0001
4014973.0005
F-2525.0001BAEA
180.91
CAD
F-2525.0001BA
F-2525.0005BA
Bz-Cit-OMe
Bz-Cit-OMe
Sequence: Bz-Cit-OMe
Bachem also offers Fmoc-Cit-Wang resin, D-1840.
Synonym(s): Bz-Orn(carbamoyl)-OMe
Formula:
C₁₄H₁₉N₃O₄ MW: 293.32 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00038142 CAS Number: 14325-36-1 EINECS: 238-273-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...