L(+)-Penicillamine

Supplier: BACHEM AMERICAS INC

Synonyms: (R)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine

4031087.0005 4031087.0025
F-2515.0005BAEA 781.5 CAD
F-2515.0005BA F-2515.0025BA
L(+)-Penicillamine
L(+)-Penicillamine

Sequence: H-Pen-OH


Corresponds to β,β-dimethylcysteine or β-mercaptovaline. For an S-protected derivative of deamino-penicillamine please see Q-1610. The rigidity of disulfide bridges can be increased by replacing Cys by Pen. Pen analogs of enkephalins, RGD-peptides, urotensin and further active peptides with improved potency and/or selectivity have been described. Bachem offers a choice of resins preloaded with Fmoc-Pen derivatives, please see the product family 'Preloaded Resins for SPPS and SPOS'.


Synonym(s): H-β,β-Dimethyl-Cys-OH#H-β-Mercapto-Val-OH

Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064303
CAS Number: 1113-41-3
EINECS: 214-203-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR