H-β-(3-Benzothienyl)-Ala-OH

Supplier: BACHEM AMERICAS INC

4014923.0001 4014923.0005
F-2490.0001BAEA 309.81 CAD
F-2490.0001BA F-2490.0005BA
H-β-(3-Benzothienyl)-Ala-OH
H-β-(3-Benzothienyl)-Ala-OH

Sequence: H-β-(3-Benzothienyl)-Ala-OH


3-Benzothienylalanine (3-Bta) is a sulfur analog of tryptophan.

Formula: C₁₁H₁₁NO₂S
Storage Temperature: Ambient
MDL Number: MFCD00079684
CAS Number: 72120-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR