H-D-Lys(boc)-OtBu·HCl
Supplier: BACHEM AMERICAS INC
Synonyms:
H-D-Lys(Boc)-OtBu.HCl, H-D-Lys(Boc)-OtBu·HCl
4012651.0001
4012651.0005
F-2455.0001BAEA
387.21
CAD
F-2455.0001BA
F-2455.0005BA
H-D-Lys(boc)-OtBu·HCl
H-D-Lys(boc)-OtBu·HCl
Sequence: H-D-Lys(Boc)-OtBu · HCl
For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
Formula:
C₁₅H₃₁ClN₂O₄ Storage Temperature: Freezer |
MDL Number:
MFCD00237143 CAS Number: 201007-86-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...