D-(-)-Citrulline
Supplier: BACHEM AMERICAS INC
Synonyms:
H-D-Cit-OH
4014198.0001
4014198.0005
F-2435.0001BAEA
258.14
CAD
F-2435.0001BA
F-2435.0005BA
D-(-)-Citrulline
D(-)-Citrulline
Sequence: H-D-Cit-OH
Bachem also offers Fmoc-Cit-Wang resin, D-1840.
Synonym(s): H-D-Orn(carbamoyl)-OH
Formula:
C₆H₁₃N₃O₃ MW: 175.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00216469 CAS Number: 13594-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...