N-Methyl-D-aspartic acid

Supplier: BACHEM AMERICAS INC

4011485.0250 4011485.1000
F-2415.0250BAEA 464.74 CAD
F-2415.0250BA F-2415.1000BA
N-Methyl-D-aspartic acid
N-Methyl-D-aspartic acid

Sequence: N-Me-D-Asp-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.
Bachem also offers the neurotransmitter NMDA, F-2145.


Synonym(s): NMDA

Formula: C₅H₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD00004226
CAS Number: 6384-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR