N-Methyl-L-tyrosine

Supplier: BACHEM AMERICAS INC

4011845.0250 4011845.1000
F-2235.0250BAEA 309.81 CAD
F-2235.0250BA F-2235.1000BA
N-Methyl-L-tyrosine
N-Methyl-L-tyrosine

Sequence: N-Me-Tyr-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₀H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD00238322
CAS Number: 537-49-5
EINECS: 208-670-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR