H-D-Lys(Z)-OMe·HCl

Supplier: BACHEM AMERICAS INC

4011834.0001 4011834.0005 4011834.0025
F-2230.0001BAEA 180.91 CAD
F-2230.0001BA F-2230.0005BA F-2230.0025BA
H-D-Lys(Z)-OMe·HCl
H-D-Lys(Z)-OMe·HCl

Sequence: H-D-Lys(Z)-OMe · HCl


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₁₅H₂₃ClN₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00077026
CAS Number: 1158-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR