D-Cysteine hydrochloride monohydrate

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, H-D-Cys-OH·HCl·H₂O, H-D-Cys-OH.HCl

4030537.0001 4030537.0005
F-2200.0001BAEA 175.66 CAD
F-2200.0001BA F-2200.0005BA
D-Cysteine hydrochloride monohydrate
D-Cysteine hydrochloride monohydrate

Sequence: H-D-Cys-OH · HCl · H₂O

Formula: C₃H₇NO₂S·HCl·H₂O
MW: 175.64 g/mol (157.62 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150051
CAS Number: 207121-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR